


Diethyl ether ≥98% (by GC) stabilised, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # BAKR9240-22
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Aluminium bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Meets ACS Reagent Chemical Requirements:≥98.0%
- For Laboratory,Research,or Manufacturing Use:≤0.001%
- Assay ((C₂H₅)₂O) (by GC, exclusive of preservative):≥98.0 %
- Carbonyl Compounds (as HCHO) (by polarography):≤0.001 %
- Color (APHA):≤10
- Residue after Evaporation:≤0.001 %
- Titrable Acid (µeq/g):≤0.2
- Peroxide (as H₂O₂):≤1 ppm
- Preservative (C₂H₅OH), For Information Only:≥0.0007%
- Preservative (H₂O), (w/w):≤0.5 %
- Preservative (BHT):≥0.0007 %
Specification Test Results
About this item
:
:74.12 g/mol
Merck Index:12,03852
Boiling Pt:35 °C (1013 hPa)
Melting Pt:–123 °C
Density:0,7135 g/cm³ (20 °C)
Flash Pt:–40 °C
Storage Temperature:Ambient
EINECS:200-467-2
UN:1155
ADR:3,I
REACH:01-2119535785-29
:GHS02
:GHS07
:GHS08
CAS Number:60-29-7
Formula:(CH₃CH₂)₂O
MDL Number:MFCD00011646