DL-Serine (not of animal origin) 98,5+% (dry basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:302-84-1
:
Formula:C₃H₇NO₃
MDL Number:MFCD00064223
:105.09 g/mol
Merck Index:14,08460
Melting Pt:∼ 240 °C (dec.)
Storage Temperature:Ambient
EINECS:206-130-6
Specification Test Results
- Assay (Dry Basis):≥98.5%
- Chloride Content:≤0.021%
- Identification (FTIR):Conforms
- Residue on Ignition:≤0.1%
- Impurity Content Sulphate (SO₄):≤0.05%
- Loss on Drying:≤0.3%
- Heavy Metals (Pb):≤20 ppm
Specification Test Results
Related Information
Organic Building Blocks and Intermediates
Application Notes
Thermo Scientific Cell Culture Workflow