Order Entry
Singapore
ContactUsLinkComponent
Spill control double-drum collection centre, EcoPolyBlend™
Spill control double-drum collection centre, EcoPolyBlend™
Catalog # JUSU28683
   Sustainable Materials
Spill control double-drum collection centre, EcoPolyBlend™
Catalog # JUSU28683
   Sustainable Materials
Specifications
  • Weight:
    49 kg
  • Description:
    Spill containment collection center, yellow, 208 liters sump capacity, for 2 drums
  • W×D×H:
    1524×991×1194 mm
  • Environmentally Preferable:
    Sustainable Materials
  • Pk:
    1
Specifications
About this item
EcoPolyBlend™ double-drum collection centre is environmentally friendly-made of recycled, durable polyethylene stands up to the harshest environments.
  • Chemical resistant
  • Dual covers can be opened separately or closed together and locked
  • Forklift channels allow for easy relocation, when empty
  • Lids include lock to prevent unauthorised access
Certifications: EPA compliant, supports SPCC, and meets NFPA 1 2009 edition and IFC fire codes.