Uricase
100 U/g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Uricase is very important for the determination of uric acid in biological fluids.
- One unit oxidises one micromole of uric acid per minute
- Form: Lyophilised powder
Uricase is an important medical enzyme which can be used to determine urate in clinical analysis. The enzyme urate oxidase (UO) catalyses the oxidation of uric acid to 5-hydroxyisourate.
Documents
Specifications
Specifications
- Pk:100 U/g
- Cat. No.:ALFAJ60875.XR
- Storage conditions:Store at –20 °C
- Enzyme synonyms:EC 1.7.3.3
- Solubility:Hardly soluble in water, slightly soluble in buffer alkaline solution
- Enzyme name:Uricase
- CAS No.:9002-12-4



