Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Hexamethyldisiloxane
Hexamethyldisiloxane, 0.65 cSt (25°C) 99.7% for NMR spectroscopy
Hexamethyldisiloxane, 0.65 cSt (25°C) 99.7% for NMR spectroscopy
:
Thermo Fisher Scientific
Hexamethyldisiloxane, 0.65 cSt (25°C) 99.7% for NMR spectroscopy
:
Thermo Fisher Scientific
:
107-46-0
:
:
C₆H₁₈OSi₂
:
MFCD00008265
:
162.38 g/mol
:
100…101 °C
:
–67 °C
:
0,764
:
–2 °C (28 °F)
:
Ambient
:
203-492-7
:
1993
:
3,II
:
01-2119496108-31
:
GHS02
msds_ALFAL16970
Thermo Scientific Acroseal 25 Years