Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Amino-5-bromo-3-methylpyridine
2-Amino-5-bromo-3-methylpyridine 97%
2-Amino-5-bromo-3-methylpyridine 97%
2-Amino-5-bromo-3-methylpyridine 97%
ALFAA15318.06
:
Thermo Fisher Scientific
:
2-Amino-5-bromo-3-methylpyridine 97%
ALFAA15318.06
:
Thermo Fisher Scientific
:
Pack type:
Glass bottle for solids
Pk:
5 g
Seq:
3173
:
3430-21-5
:
:
C₆H₇BrN₂
:
MFCD00068232
:
187.04 g/mol
:
90…94 °C
:
GHS07
:
Ambient