$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Singapore
ContactUsLinkComponent
1H-Pyrazole
1H-Pyrazole ≥98%
1H-Pyrazole ≥98%
:
Thermo Fisher Scientific
1H-Pyrazole ≥98%
:
Thermo Fisher Scientific
:
288-13-1
:
:
C₃H₄N₂
:
MFCD00005234
:
14,7960
:
186…188 °C
:
66…70 °C
:
1,34 g/cm³ (20 °C)
:
68.08 g/mol
:
Ambient
:
206-017-1
:
GHS08
:
GHS07
msds_ALFAA14186
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years