


Cobalt(II) chloride, ultra dry ≥99.9% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7646-79-9
:
Formula:Cl₂Co
MDL Number:MFCD00010938
:129.84 g/mol
Merck Index:14,2437
Boiling Pt:1049 °C
Melting Pt:735 °C
Density:3,367
Storage Temperature:Ambient
UN:3077
ADR:9,III
:ampouled under argon
EINECS:231-589-4
:GHS08
:GHS07
:GHS09