Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Thulium (III) oxide
Thulium(III) oxide ≥99.997% (REO, rare earth oxide basis), REacton®
Thulium(III) oxide ≥99.997% (REO, rare earth oxide basis), REacton®
:
Thermo Fisher Scientific
Thulium(III) oxide ≥99.997% (REO, rare earth oxide basis), REacton®
:
Thermo Fisher Scientific
:
12036-44-1
:
:
O₃Tm₂
:
MFCD00011285
:
385.87 g/mol
:
14,9394
:
2425 °C
:
8,6 g/cm³ (20 °C)
:
234-851-6
:
GHS07
:
Ambient
msds_ALFA010918
Thermo Scientific Acroseal 25 Years