Order Entry
Singapore
ContactUsLinkComponent
 
Ammonium sulphate ≥99.5%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
 
undefined
Ammonium sulphate ≥99.5%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
 

 

  • Assay:
    Min. 99.5 %
  • pH (25°C; 5 %):
    5.0 - 6.0
  • Heavy metals (as Pb):
    Max. 5 ppm
  • Ignition residue (SO4):
    Max. 50 ppm
  • Insolubility in water:
    Max. 10 ppm
  • Loss on drying (105°C):
    Max. 0.1 %
  • Cl (Chloride):
    Max. 3 ppm
  • NO3 (Nitrate):
    Max. 10 ppm
  • PO4 (Phosphate):
    Max. 5 ppm
  • As (Arsenic):
    Max. 0.2 ppm
  • Ca (Calcium):
    Max. 10 ppm
  • Cd (Cadmium):
    Max. 1 ppm
  • Cu (Copper):
    Max. 2 ppm
  • Fe (Iron):
    Max. 2 ppm
  • K (Potassium):
    Max. 20 ppm
  • Mg (Magnesium):
    Max. 5 ppm
  • Na (Sodium):
    Max. 20 ppm
  • Pb (Lead):
    Max. 2 ppm
  • Zn (Zinc):
    Max. 1 ppm
  • Conforms to ACS:
    Passes test
  • Conforms to Reag. Ph.Eur.:
    Passes test