Order Entry
Singapore
Orders LinkContactUsLinkComponent
Cetrimonium bromide

Cetrimonium bromide

Plastic bottle for solids, 500 g
Catalog # APLIA6284.0500
Health
Irritant
Corrosive
Environment
Supplier:   PanReac AppliChem
Cetrimonium bromide
Catalog # APLIA6284.0500
Supplier:  PanReac AppliChem
CAS Number:  57-09-0
Cetrimonium bromide

Plastic bottle for solids, 500 g

CAS Number:  57-09-0
Price
$2,200.00
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  N-Cetyl-N,N,N-trimethylammonium bromide, Cetyltrimethylammonium bromide, Hexadecyltrimethylammonium bromide, CTAB, (1-Hexadecyl)trimethylammonium bromide, N,N,N-Trimethylhexadecan-1-aminium bromide
  • CAS Number:57-09-0
  • Formula:CH₃(CH₂)₁₅N(Br)(CH₃)₃
  • MDL Number:MFCD00011772
  • Molecular Weight:364.45 g/mol
  • Merck Index:13,02034
  • Melting Pt:237…243 °C
  • Flash Pt:244 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:200-311-3
  • UN:3077
  • ADR:9,III
Specifications

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    500 g

Specifications