Tributyl phosphate, J.T.Baker®
BAKRW432-07
: Avantor
:
- Pack type:Glass bottle
- Pk:500 mL
- Seq:2725
- For Laboratory,Research,or Manufacturing Use:Passes Test
- Identification (by IR):Passes Test
- Refractive Index, n²⁵^D:1.4205 - 1.4225
:
:266.32 g/mol
:13,09692
:289 °C (1013 hPa)
:–79 °C
:0,982 g/cm³ (20 °C)
:151 °C
:204-800-2
:GHS08
:GHS09
:Ambient
:126-73-8
:(CH₃(CH₂)₃O)₃PO
:MFCD00009436