Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Tin (II) oxide
Tin(II) oxide ≥99%
Tin(II) oxide ≥99%
:
Thermo Fisher Scientific
Tin(II) oxide ≥99%
:
Thermo Fisher Scientific
:
21651-19-4
:
:
OSn
:
MFCD00011243
:
134.71 g/mol
:
14,8787
:
1425 °C (1013 hPa)
:
1080 °C (dec.)
:
6,45
:
244-499-5
:
GHS07
:
Ambient
msds_ALFAA17889
Thermo Scientific Acroseal 25 Years