Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Lead (II) iodide
Lead(II) iodide ≥99.9985% (metals basis)
Lead(II) iodide ≥99.9985% (metals basis)
:
Thermo Fisher Scientific
Lead(II) iodide ≥99.9985% (metals basis)
:
Thermo Fisher Scientific
:
10101-63-0
:
:
I₂Pb
:
MFCD00011163
:
14,5411
:
954 °C
:
402 °C
:
6,16
:
Ambient
:
233-256-9
:
6.1,III
:
461.01 g/mol
:
2291
:
GHS08
:
GHS07
:
GHS09
externalurl.pdf
msds_ALFA012724
Thermo Scientific Acroseal 25 Years