Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Magnesium oxide
Magnesium oxide ≥99.95% (metals basis)
Magnesium oxide ≥99.95% (metals basis)
:
Thermo Fisher Scientific
Magnesium oxide ≥99.95% (metals basis)
:
Thermo Fisher Scientific
:
Magnesia usta
:
1309-48-4
:
:
MgO
:
MFCD00011109
:
40.3 g/mol
:
14,5677
:
3600 °C
:
2852 °C
:
3,58
:
Ambient
:
215-171-9
Environmentally Preferable:
msds_ALFA043196
Thermo Scientific Acroseal 25 Years