Order Entry
Singapore
ContactUsLinkComponent
Magnesium carbonate 40,0 - 43,5%, powder USP, FCC, Macron Fine Chemicals™
Magnesium carbonate 40,0 - 43,5%, powder USP, FCC, Macron Fine Chemicals™
Catalog # MACR5942-32
CAS Number:  
undefined
Magnesium carbonate 40,0 - 43,5%, powder USP, FCC, Macron Fine Chemicals™
Catalog # MACR5942-32
CAS Number:  

Specifications

  • Pack type:
    Fiber drum, Leverpack®
  • Pk:
    50 lb
  • Seq:
    7137
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Food GMP Manufactured Product:
  • Meets F.C.C. Requirements:
  • GMP Manufactured Product:
  • Meets U.S.P Requirements:
  • CAUTION: For Manufacturing, processing or repackaging:
  • Bulk Pharmaceutical Chemical:
  • Bulk Food Chemical:
  • USP - Assay (MgO):
    40.0 - 43.5 %
  • USP - Identification A:
    Passes Test
  • USP - Identification B:
    Passes Test
  • USP - Identification C:
    Passes Test
  • USP - Microbiological:
    Passes Test
  • USP - Soluble Salts:
    ≤ 1.0 %
  • USP - Acid-Insoluble Substances:
    ≤ 0.05 %
  • USP - Arsenic (As):
    ≤ 3 ppm
  • USP - Limit of Calcium (Ca):
    ≤ 0.45 %
  • USP - Iron (Fe):
    ≤ 0.02 %
  • FCC - Assay (MgO):
    40.0 - 43.5 %
  • FCC - Identification:
    Passes Test
  • FCC - Soluble Salts:
    ≤ 1.0 %
  • FCC - Acid-Insoluble Substances:
    ≤ 0.05 %
  • FCC - Lead (Pb):
    ≤ 2 mg/kg
  • FCC - Calcium Oxide:
    ≤ 0.6 %
  • Appearance (bulky, white powder):
    Passes Test
  • Solubility in Dilute Sulfuric Acid:
    Passes Test
  • Residue on US 325 Mesh:
    ≤ 0.3 %
  • Tapped Density:
    ≥ 0.43 g/ml

Specification Test Results

About this item

Synonyms: Magnesium Carbonate
 :
 :84.31 g/mol
EINECS:208-915-9
CAS Number:546-93-0
Formula:CMgO₃
MDL Number:MFCD00064632
Density:3.03 g/cm³ (16 °C)
 :Management of Change (MOC) category = R