$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Singapore
ContactUsLinkComponent
Potassium dihydrogen phosphate
Potassium dihydrogen phosphate ≥99.0% ACS
Potassium dihydrogen phosphate ≥99.0% ACS
:
Thermo Fisher Scientific
Potassium dihydrogen phosphate ≥99.0% ACS
:
Thermo Fisher Scientific
:
7778-77-0
:
:
KH₂PO₄
:
MFCD00011401
:
136.09 g/mol
:
14,7659
:
∼100 °C (1013 hPa)
:
253 °C
:
2,338
:
Ambient
:
231-913-4
:
01-2119490224-41
:
GHS07
ALFA11594.30_TDS_EN
msds_ALFA011594
Thermo Scientific Acroseal 25 Years