

Potassium hydroxide 85.0-100.5%, pellets Ph. Eur.
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:1310-58-3
:
Formula:KOH
MDL Number:MFCD00003553
:56.11 g/mol
Merck Index:12,07806
Boiling Pt:1320 °C (1013 hPa)
Melting Pt:360 °C
Density:2,04 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-181-3
UN:1813
ADR:8,II
REACH:01-2119487136-33
:GHS07
:GHS05
Specification Test Results
- Assay (calculated as KOH):85.0 - 100.5 %
- Appearance:Conforms (see CoA/CoS)
- pH (20°C; 0,01 %):Min. 10.5
- Identification B (K):Passes test
- Solution S1:Passes test
- Solution S2:Passes test
- Appearance of solution:Passes test
- CO3 (as K2CO3):Max. 2.0 %
- Cl (Chloride):Max. 200 ppm
- PO4 (Phosphate):Max. 100 ppm
- SO4 (Sulphate):Max. 200 ppm
- Fe (Iron):Max. 10 ppm
- Na (Sodium):Max. 1.0 %
- Heavy metals (as Pb):Max. 10 ppm
- Residual solvents:Passes test
- Conforms to Ph.Eur.:Passes test
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Plastic bottle for solids DIN 60 UN (ZPACK13251000) 6L Plastic bottle for solids DIN 80 (ZPACK13256000)
Packaging Information
BSE_26670_POTASSIUM HYDROXIDE PELLETS PH.EUR
BSE/TSE Certificates
Potassium Hydroxide 85 - 100,5%, Pellets Ph. Eur.
Additional Product Details
spec_VWRC26670.294 spec_VWRC26670.294 spec_VWRC26670.294 spec_VWRC26670.363 spec_VWRC26670.363
Product specification
More
VWR Chemicals for Biopharma and Pharma Production