

Potassium periodate 99.8-100.3% (by iodometry), BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:7790-21-8
:
Formula:KlO₄
MDL Number:MFCD00011407
:230 g/mol
Merck Index:13,07739
Melting Pt:576…588 °C
Density:3,62 g/cm³ (25 °C)
EINECS:232-196-0
UN:1479
ADR:5.1,II
:GHS03
:GHS07
Specification Test Results
- Assay (KIO₄):99.8 - 100.3 %
- Insoluble Matter:≤0.01 %
- Other Halogens (as Cl):≤0.01 %
- Trace Impurities - Manganese (Mn):≤0.500 ppm