Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Bromo-2,6-xylidine
4-Bromo-2,6-xylidine ≥98%
4-Bromo-2,6-xylidine ≥98%
:
Thermo Fisher Scientific
4-Bromo-2,6-xylidine ≥98%
:
Thermo Fisher Scientific
:
24596-19-8
:
:
C₈H₁₀BrN
:
MFCD00007826
:
120 °C (0,2 mmHg)
:
47…52 °C
:
>110 °C (230 °F)
:
Ambient
:
246-337-9
:
2811
:
6.1,III
:
200.08 g/mol
:
GHS06
:
GHS08
:
GHS07
msds_ALFAA10712
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years