Bromocresol green
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:76-60-8
:
Formula:C₂₁H₁₄Br₄O₅S
MDL Number:MFCD00005874
:698.02 g/mol
Merck Index:14,1386
Melting Pt:∼ 225 °C (dec.)
Storage Temperature:Ambient
EINECS:200-972-8