3-Nitro-o-cresol 96%
Catalog # ALFAA13002.14
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:153.14 g/mol
Melting Pt:148…152 °C
Storage Temperature:Ambient
EINECS:226-739-0
UN:2446
ADR:6.1,III
:GHS06
:GHS07
CAS Number:5460-31-1
Formula:C₇H₇NO₃
MDL Number:MFCD00007241