

4-Nitro-2-(trifluoromethyl)aniline ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:121-01-7
:
Formula:C₇H₅F₃N₂O₂
MDL Number:MFCD00007365
Melting Pt:90…93 °C
Density:1,754 g/cm³ (–123.15 °C)
Storage Temperature:Ambient
EINECS:204-443-2
UN:2811
ADR:6.1,III
:206.12 g/mol
:GHS06
:GHS07