Haematoxylin solution (Mayer)
1
ELMI26043-05
Marketsource
- Stain selectivity for nuclei
- High quality staining solution
- Ready-to-use
: DcH-48, Natural Black I, Hydroxybrasilin
- :517-28-2
- :C₁₆H₁₄O₆
- :MFCD00078111
- :302.28 g/mol
- :96 °C (205 °F)
- :Ambient
- Pk:1