Osmium tetraoxide ≥99.95%, crystalline, highest purity
Catalog # ELMI19102
Supplier: ELECTRON MICROSCOPY SCIENCES
CAS Number:
Specifications
- Pack type:Glass Ampoule
- Pk:20 g
- Seq:3290
- Pk Info:20x1 g
Specifications
Specification Test Results
- Vapor pressure at 25 °C:9.8 mmHg
- Osmium purity:>99.95%
- Non-Volatile matter:<0.02%
Specification Test Results
About this item
:
:254.23 g/mol
Boiling Pt:130 °C
Melting Pt:40,1 °C
Density:4,906 g/cm³ (22°C)
UN:2471
ADR:6.1,I
:GHS05
:GHS06
Storage Temperature:Ambient
CAS Number:20816-12-0
Formula:O₄Os
MDL Number:MFCD00011150