Mucicarmine stock solution
1
Catalog # ELMI26080-05
Marketsource
Supplier: ELECTRON MICROSCOPY SCIENCES
Mucicarmine stock solution
Catalog # ELMI26080-05
Supplier: ELECTRON MICROSCOPY SCIENCES
Marketsource
CAS Number: 51395-97-2
Jump to:
Product Details & Documents
Mucicarmine is for use with hematoxylin and metanil yellow for the determination of carbohydrates and mucosubstances.
- CAS Number:51395-97-2
- MDL Number:MFCD00071289
Specifications
Specifications
- Pk:1



