Order Entry
Singapore
ContactUsLinkComponent
Glutaraldehyde 25% in aqueous solution, EM Grade
Glutaraldehyde 25% in aqueous solution, EM Grade
Catalog # ELMI16220
CAS Number:  
Glutaraldehyde 25% in aqueous solution, EM Grade
Catalog # ELMI16220
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Seq:
    2440
  • Pk Info:
    10× 10 ampoules

Specifications

About this item

 :
 :100.12 g/mol
UN:3265
ADR:8,II
 :GHS05
 :GHS06
 :GHS08
CAS Number:111-30-8
Formula:C₅H₈O₂
MDL Number:MFCD00007025