




Formaldehyde 35,5 -38,0% stabilised ACS
Catalog # ELMI15680
Supplier: ELECTRON MICROSCOPY SCIENCES
CAS Number:
Specifications
- Pack type:Bottle
- Pk:450 mL
- Seq:2675
Specifications
Specification Test Results
- Assay:35.5 to 38.0%
- Residue after ignition:0.005%
- Color (APHA):10
- Sulfate:0.002%
- Heavy Metal (as Pb):5ppm
- Titratable acid:0.006 meq/g
- Chloride (Cl):5ppm
- Iron (Fe):5ppm
Specification Test Results
About this item
:
:30.03 g/mol
Boiling Pt:64 °C (147 °F)
Density:1,09 g/ml
Flash Pt:60 °C (140 °F
Storage Temperature:Ambient
UN:1198
ADR:3,III
:Stabilised with methanol
:GHS02
:GHS05
:GHS06
:GHS07
:GHS08
CAS Number:50-00-0
Formula:CH₂O
MDL Number:MFCD00003274