Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Bromo-p-cresol
3-Bromo-p-cresol ≥98%
3-Bromo-p-cresol ≥98%
3-Bromo-p-cresol ≥98%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
3-Bromo-p-cresol ≥98%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
60710-39-6
:
Formula:
C₇H₇BrO
MDL Number:
MFCD08273793
:
187.04 g/mol
Melting Pt:
54…57 °C
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAH28689
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation