Phase separating filters
Supplier: Advantech
About this item
Separate aqueous and non-aqueous phases of mixtures. Hydrophobic filters retain aqueous phase while non-aqueous phase passes through.
These filters are made from silicone treated cellulose.
3 Options Available Below
Product Details & Documents
Separate aqueous and non-aqueous phases of mixtures. Hydrophobic filters retain aqueous phase while non-aqueous phase passes through.
These filters are made from silicone treated cellulose.
Specifications for Product Family
Specifications
- Catalog No:
- ADVTN02S12.5CM
- ADVTN02S15.0CM
- ADVTN02S18.5CM
- Weight:
- 120 g/m²
- 120 g/m²
- 120 g/m²
- Thickness:
- 0,26 mm
- 0,26 mm
- 0,26 mm
Specifications
Product Family Options
Product Information
- GradeSizeAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.ADVTN02S12.5CMWeight120 g/m²Thickness0,26 mm - Specifications:
More
Specifications
Cat. No.ADVTN02S15.0CMWeight120 g/m²Thickness0,26 mm - Specifications:
More
Specifications
Cat. No.ADVTN02S18.5CMWeight120 g/m²Thickness0,26 mm