Order Entry
Singapore
Orders LinkContactUsLinkComponent
Filter paper for blood sampling
Filter paper for blood sampling
Catalog # ADVT49010010
Supplier:   Advantech
Filter paper for blood sampling
Catalog # ADVT49010010
Supplier:  Advantech
Filter paper for blood sampling
Price
Inquire For Price
Log in for availability
1 * 100 items
Product Details & Documents
These blood filter strips are used to track healthy and diseased animal populations safely, efficiently, and economically.
  • High purity cellulose fibers for consistent, uniform sampling
  • Each strip holds approximately 1 ml of blood or 0,04 ml serum
  • Filter strips require no refrigeration or pretreatment for sampling, storage, and analysis
Collect blood on the narrow end of the strip and allow to dry on the convenient drying rack for easy and efficient sample handling.