ω-Bromo-4-chloroacetophenone ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:536-38-9
:
Formula:C₈H₆BrClO
MDL Number:MFCD00000625
:233.49 g/mol
Merck Index:14,2153
Boiling Pt:278 °C (1013 hPa)
Melting Pt:94…98 °C
Storage Temperature:Ambient
EINECS:208-631-5
UN:3261
ADR:8,III
:GHS05