5-Bromo-2-chlorotoluene ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:54932-72-8
:
Formula:C₇H₆BrCl
MDL Number:MFCD00018503
:205.48 g/mol
Boiling Pt:90…93 °C (10 mmHg)
Density:1,549
Flash Pt:96 °C (205 °F)
:GHS07
Storage Temperature:Ambient