1-Bromo-4-hexylbenzene ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:23703-22-2
:
Formula:C₁₂H₁₇Br
MDL Number:MFCD00061114
:241.17 g/mol
Boiling Pt:280 °C
Density:1,179
Flash Pt:>120 °C (248 °F)
Storage Temperature:Ambient