Order Entry
Singapore
ContactUsLinkComponent
 
Potassium hydrogen phthalate ≥99.5%, AnalaR NORMAPUR® analytical reagent
 
undefined
Potassium hydrogen phthalate ≥99.5%, AnalaR NORMAPUR® analytical reagent
 

 

  • Assay (calculated on dried substance):
    Min. 99.5 %
  • IR Spectrum:
    Passes test
  • Heavy metals (as Pb):
    Max. 5 ppm
  • Insolubility in water:
    Max. 50 ppm
  • Loss on drying (105°C):
    Max. 0.2 %
  • Sulphur compounds (as SO4):
    Max. 50 ppm
  • Cl (Chloride):
    Max. 20 ppm
  • Cu (Copper):
    Max. 2 ppm
  • Fe (Iron):
    Max. 5 ppm
  • Na (Sodium):
    Max. 100 ppm
  • Pb (Lead):
    Max. 5 ppm