Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,4-Dichloro-5-nitrophenol
2,4-Dichloro-5-nitrophenol ≥98%
2,4-Dichloro-5-nitrophenol ≥98%
2,4-Dichloro-5-nitrophenol ≥98%
ALFAL18369.14
:
Thermo Fisher Scientific
:
2,4-Dichloro-5-nitrophenol ≥98%
ALFAL18369.14
:
Thermo Fisher Scientific
:
Pack type:
Glass bottle for solids
Pk:
25 g
Seq:
3308
:
:
208 g/mol
:
97…102 °C
:
Ambient
:
2811
:
6.1,III
:
GHS06
:
GHS07
:
39489-77-5
:
C₆H₃Cl₂NO₃
:
MFCD03093072