4-Bromo-2-fluorobenzoyl chloride ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:151982-51-3
:
Formula:C₇H₃BrClFO
MDL Number:MFCD03411584
:237.46 g/mol
Boiling Pt:86…88 °C (15 mmHg)
Melting Pt:24…25 °C
Storage Temperature:Ambient
UN:3261
ADR:8,II
:GHS05