Ethyl-4-bromobenzoate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:5798-75-4
:
Formula:C₉H₉BrO₂
MDL Number:MFCD00016329
:229.07 g/mol
Merck Index:14,1408
Boiling Pt:262…264 °C
Melting Pt:18 °C
Density:1,403
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
EINECS:227-347-2