2-Bromo-4,6-dichloroaniline ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:697-86-9
:
Formula:C₆H₄BrCl₂N
MDL Number:MFCD00040936
:240.91 g/mol
Boiling Pt:273 °C (1013 hPa)
Melting Pt:78…82 °C
:GHS06
:GHS07
Storage Temperature:Ambient