4-Hydroxy-1-methylpiperidine ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:106-52-5
:
Formula:C₆H₁₃NO
MDL Number:MFCD00006500
:115.18 g/mol
Boiling Pt:198…200 °C
Melting Pt:26…30 °C
Density:0,98
Flash Pt:87 °C (188 °F)
EINECS:203-406-8
:GHS07
Storage Temperature:Ambient