


1-Ethyl-2-pyrrolidone ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2687-91-4
:
Formula:C₆H₁₁NO
MDL Number:MFCD00003199
:113.16 g/mol
Boiling Pt:96…98 °C (20 mmHg)
Density:0,992
Flash Pt:76 °C (169 °F)
EINECS:220-250-6
:GHS08
:GHS05
:GHS07
Storage Temperature:Ambient