Order Entry
Singapore
Orders LinkContactUsLinkComponent
Test papers, ion
Test papers, ion
 :   Advantech
Test papers, ion
 :  Advantech
Test papers, ion
Test papers, ion
Detects concentration of specified ion in solution. Two-point indicator strip confirms within pH range and ion concentration by color comparison.
Dip the indicator test strip in solution and compare to the color chart on the container.
Packaging: Strips, 5×85 mm.
Product Family Options
  • Description
    pH range
    Range
  • Aluminium Check (Al3+)
    3.0 – 6.0
    0 − 50 mg/l
    1 * 100 items
    1 * 100 items
  • Copper Check (Cu)
    3.0 – 5.0
    0 – 1000 mg/l
    1 * 100 items
    1 * 100 items
  • Copper Check-A (Cu+, Cu2+)
    1.0 – 6.0
    0 – 1000 mg/l
    1 * 100 items
    1 * 100 items
  • Iron Check (Fe2+)
    1.0 – 6.0
    0 – 500 mg/l
    1 * 100 items
    1 * 100 items
  • Nickel Check (Ni2+)
    2.0 – 7.0
    0 − 50 mg/l
    1 * 100 items
    1 * 100 items
  • Cyanogen Check (CN-)
    7.0 – 9.0
    0 – 200 mg/l
    1 * 100 items
    1 * 100 items
  • Chrome Check (Cr6+)
    1.0 – 4.0
    0 – 1000 mg/l
    1 * 25 items
    1 * 25 items
  • Chrome Check-A (Cr6+)
    1.0 – 9.0
    0 – 100 mg/l
    1 * 25 items
    1 * 25 items