Ethyl isothiocyanate ≥97%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 97% CAS No.: 542-85-8 Molecular Formula: CH3CH2NCS Formula Weight: 87.14 Flash Point: 32°(89°F) Refractive Index: 1.5110 Odour: Pungent Melting Point: -6° Boiling Point: 130-132° Beilstein Reference: 1737705 Merck Reference: 14,3815 UN No.: 2929 MDL No.: MFCD00004820
- CAS Number:542-85-8
- Formula:C₃H₅NS
- MDL Number:MFCD00004820
- Molecular Weight:87.15 g/mol
- Merck Index:14,3815
- Boiling Pt:130…132 °C
- Melting Pt:–6 °C
- Density:0,995
- Flash Pt:32 °C (89 °F)
- Storage Temperature:Ambient
- EINECS:208-831-2
- UN:2929
- ADR:6.1,II
3 Options Available Below
Product Details & Documents
Purity: 97% CAS No.: 542-85-8 Molecular Formula: CH3CH2NCS Formula Weight: 87.14 Flash Point: 32°(89°F) Refractive Index: 1.5110 Odour: Pungent Melting Point: -6° Boiling Point: 130-132° Beilstein Reference: 1737705 Merck Reference: 14,3815 UN No.: 2929 MDL No.: MFCD00004820
- CAS Number:542-85-8
- Formula:C₃H₅NS
- MDL Number:MFCD00004820
- Molecular Weight:87.15 g/mol
- Merck Index:14,3815
- Boiling Pt:130…132 °C
- Melting Pt:–6 °C
- Density:0,995
- Flash Pt:32 °C (89 °F)
- Storage Temperature:Ambient
- EINECS:208-831-2
- UN:2929
- ADR:6.1,II
Documents
Product Family Options
Product Information
- Pack typePkAvailabilityPrice



