
Ethyl bromopyruvate ≥75%, Technical Grade
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:70-23-5
:
Formula:BrH₂CC(O)COOCH₂CH₃
MDL Number:MFCD00000204
:195.01 g/mol
Boiling Pt:98…100 °C (10 mmHg)
Density:1,555
Flash Pt:98 °C (208 °F)
Storage Temperature:Refrigerator
EINECS:200-729-6
UN:3265
ADR:8,III
:GHS05