

1-Phenyl-2-tetrazoline-5-thione ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:86-93-1
:
Formula:C₇H₆N₄S
MDL Number:MFCD00003129
:178.22 g/mol
Melting Pt:∼ 145 °C (dec.)
Flash Pt:138 °C (280 °F)
Storage Temperature:Ambient
EINECS:201-710-5
UN:1325
ADR:4.1,III
:GHS02
:GHS07