D-(-)-Sorbitol
Glass bottle for solids, 1,2 g
Catalog # EDQMS1000000
Supplier: EDQM
Jump to:
Product Details & Documents
Synonyms: (2R,3R,4R,5S)-Hexane-1,2,3,4,5,6-hexaol, (Sorbitol), D(-)-Glucitol, D-Sorbitol, Sorbitol
- CAS Number:50-70-4
- Formula:HOH₂C(CH(OH))₄CH₂OH
- MDL Number:MFCD00004708
- Molecular Weight:182.17 g/mol
- Merck Index:13,08797
- Boiling Pt:295 °C (1013 hPa)
- Melting Pt:94…98 °C
- Density:1,524 g/cm³ (20 °C)
- Flash Pt:>100 °C
- Storage Temperature:Ambient
- EINECS:200-061-5
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1,2 g



