L(+)-Glutamic acid 98.5-101.5%, high purity
Catalog # VWRC0421-1KG
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
Specification Test Results
- Heavy Metals (as Pb):< = 0.002 %
- Lead:<= 0.001 %
- Loss on Drying:<= 0.1 %
- Purity (Dried):98.5 % - 101.5 %
- Specific Rotation (Dried):31.5 ° - 32.5 °
Specification Test Results
About this item
:
:147.13 g/mol
Merck Index:13,04482
Boiling Pt:325 °C (1013 hPa)
Melting Pt:160 °C
Density:1,538 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-293-7
CAS Number:56-86-0
Formula:HO₂CCH₂CH₂CH(NH₂)CO₂H
MDL Number:MFCD00002634