

Iron(II) chloride tetrahydrate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13478-10-9
:
Formula:FeCl₂·4H₂O
MDL Number:MFCD00149709
Merck Index:14,4043
Melting Pt:677 °C
Density:1,93
EINECS:231-843-4
UN:3260
ADR:8,III
:198.81 g/mol
:GHS05
:GHS07
Storage Temperature:Ambient