Order Entry
Singapore
ContactUsLinkComponent
Copper(II) fluoride dihydrate ≥99.99% (metals basis), Puratronic®
Corrosive
Irritant
Copper(II) fluoride dihydrate ≥99.99% (metals basis), Puratronic®
Catalog # ALFA13001.04
CAS Number:  
Corrosive
Irritant
Copper(II) fluoride dihydrate ≥99.99% (metals basis), Puratronic®
Catalog # ALFA13001.04
CAS Number:  

Specifications

  • Pk:
    2 g
  • Seq:
    3107

Specifications

About this item

 :
 :137.57 g/mol
Merck Index:14,2638
Melting Pt:>130 °C (dec.)
Density:2,934
Storage Temperature:Ambient
EINECS:232-147-3
UN:3260
ADR:8,III
 :GHS05
 :GHS07
CAS Number:13454-88-1
Formula:CuF₂·2H₂O
MDL Number:MFCD00010977