Order Entry
Singapore
ContactUsLinkComponent
 
Aluminium potassium sulphate dodecahydrate ≥99.5%, Puriss. p.a. ACS, Reag. Ph. Eur., Fluka™
 
Aluminium potassium sulphate dodecahydrate ≥99.5%, Puriss. p.a. ACS, Reag. Ph. Eur., Fluka™

 

  • Assay:
    Min. 99.5 %
  • Water insoluble matter:
    Max. 0.005 %
  • pH (10 %, 20°C):
    3.0 - 3.5
  • Ammonium (NH₄):
    Max. 0.005 %
  • Arsenic (As):
    Max. 0.0002 %
  • Cadmium (Cd):
    Max. 0.0005 %
  • Copper (Cu):
    Max. 0.0005 %
  • Iron (Fe):
    Max. 5 ppm
  • Sodium (Na):
    Max. 0.01 %
  • Lead (Pb):
    Max. 0.001 %
  • Heavy metals (as Pb):
    Max. 0.001 %
  • Chloride (Cl):
    Max. 0.0005 %
  • Appearance of the solution:
    complying